Potassium iodide, Suprapur® Trace_metal, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Dipotassium diiodide, Kali iodide

1.05044.0050 1.05044.0500
CA1.05044.0050EA 305 CAD
CA1.05044.0050 CA1.05044.0500
Potassium iodide, Suprapur® Trace_metal, Supelco®
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Solubility 1430 g/l (20 °C)
Melting Point 723 °C
Molar Mass 166.00 g/mol
Bulk Density 1500 kg/m3
Boiling Point 1325 °C (1013 hPa)
Vapor Pressure 1.3 hPa (745 °C)
Density 3.13 g/cm3 (20 °C)
pH 6.9 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR