Potassium iodide 99,5-100,5 % silver nitrate_dry [≥99.5% (silver nitrate titration)], EMSURE® analysis_for, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Dipotassium diiodide, Kali iodide

1.05043.0250 1.05043.0500 1.05043.1000 1.05043.2500 1.05043.9050
CA1.05043.0250EA 318 CAD
CA1.05043.0250 CA1.05043.0500 CA1.05043.1000 CA1.05043.2500 CA1.05043.9050
Potassium iodide 99,5-100,5 % silver nitrate_dry [≥99.5% (silver nitrate titration)], EMSURE® analysis_for, Supelco®
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686 (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Solubility 1430 g/l (20 °C)
Melting Point 723 °C
Molar Mass 166.00 g/mol
Bulk Density 1500 kg/m3
Boiling Point 1325 °C (1013 hPa)
Vapor Pressure 1.3 hPa (745 °C)
Density 3.13 g/cm3 (20 °C)
pH 6.9 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR