5' DNA Adenylation Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E2610S E2610L
CA101641-560EA 204 CAD
CA101641-560 CA102500-004
5' DNA Adenylation Kit, New England Biolabs
Assays Nucleic Acid Assays
The 5´ DNA adenylation Kit is a simple and efficient enzymatic method for generating 5´-adenylated DNA
The kit is optimized to produce the adenylated DNA with or without 3´-terminator. The 5´ DNA adenylation kit routinely generates greater than 95% conversion of pDNA to AppDNA. This highly efficient process eliminates the need for gel isolation of the product and increases overall yield.