Amersham ECL Plex Western Blotting Combination Packs, Cytiva
Supplier: Cytiva
ECL™
RPN998 RPN999 RPN999 RPN998
CA95040-084LEA 645.27 CAD
CA95040-084L CA95040-086L 95040-084 95040-086
Amersham ECL Plex Western Blotting Combination Packs, Cytiva
Electrophoresis Reagents Western Blotting Reagents
Amersham ECL Plex™ Western blotting combination packs offer a complete system for fluorescent Western blotting, with improved formulation Cy3 conjugates for Lower background.
Delivery information: Combination packs include ECL Plex goat mouse IgG-Cy3, 150 µg; ECL Plex goat rabbit IgG-Cy5, 150 µg; ECL Plex Fluorescent Rainbow Markers, 120 µl; Hybond ECL, 10×10 cm, 10 sheets or Hybond-LFP, 20×20 cm, 3 sheets Protocol, sufficient for at least 1000 cm² of membrane.