Tryptic Soy Broth (TSB)
Supplier: HARDY DIAGNOSTICS
U67
U141
U133
U71
U42
U46
K82
U82
R36
K380
U38
U223
R31
R41
U267
U44
U205
K89
R30
U70
K126
U127
K83
U367
U274
U421
U435
89407-988EA
772.97
CAD
89407-988
CA89407-994
CA89407-996
CA89408-044
CA89408-046
CA89408-048
CA89408-050
CA89408-052
CA89408-060
CA89408-062
CA89408-066
CA89408-068
CA89408-072
CA89408-076
CA89408-078
CA89408-080
CA89408-084
CA89408-086
CA89408-092
CA89408-094
CA89408-096
CA89408-100
CA89408-102
CA89408-108
CA89408-110
CA89408-112
CA89408-122
CA89408-124
CA10127-496
CA75873-762
CA89408-089
CA10124-504
CA10124-512
Tryptic Soy Broth (TSB)
Media for Microbiology
Prepared Media for Microbiology
For the general cultivation of microorganisms.
Tryptic Soy Broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi.
Delivery information: Ships ground only.