Agarose LE, Molecular Biology Grade, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C8740 C8741
CA89406-784EA 215 CAD
CA89406-784 CA89406-786
Agarose LE, Molecular Biology Grade, Hardy Diagnostics
Electrophoresis Gels Agarose and Acrylamide
CulGenex™ Agarose LE, Molecular Biology Grade is recommended for use in the preparation of an electrophoresis gel for the analysis of nucleic acids and proteins
  • Ability to vary the pore size by modifying the gel concentration
  • Easy preparation of the gel by simple dilution in aqueous buffers either by standard boiling or microwaving
  • Greater thermal stability due to high hysteresis (difference between gelling and melting temperatures)
  • Excellent transparency and exceptionally low absorption of staining agents
  • Absence of toxicity (polyacrylamide is neurotoxic)
More Product Information