GFP-CERTIFIED® FLUOFORTE® Reagent, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-52016-5C50 ENZ-52016-M001
89165-962EA 538.78 CAD
89165-962 89165-964
GFP-CERTIFIED® FLUOFORTE® Reagent, Enzo Life Sciences
Protein Linking and Labeling Systems
Calcium dye.
The GFP-Certified™ FluoForte™ Reagent is the brightest and most sensitive fluorescent calcium indictor relative to other commercially available red fluorescent dyes with maximum excitation at ~530 nm and maximum emission at ~555 nm. This product is used in a homogeneous fluorescence-based assay for detecting intracellular calcium mobilization across a broad spectrum of biological targets.