Calcein AM ≥95% (by HPLC), Ultrapure

Supplier: Enzo Life Sciences

Synonyms: Calcein-AMCalcein O,O′-diacetate tetrakis(acetoxymethyl) esterCalcein acetoxymethyl esterTetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate

ENZ-52002
89165-934EA 279 CAD
89165-934
Calcein AM ≥95% (by HPLC), Ultrapure
Calcein AM

Calcium dye


Calcein acetomethoxy (AM), a derivative of calcein, is useful for differentiating between live and dead cells. Calcein AM readily passes through the cellular plasma membrane. Once inside, esterases cleave the AM groups yielding the more hydrophilic calcein, trapped inside the cell. The loss of the acetomethoxy group also enables calcein to readily bind intracellular calcium; resulting in a strong yellowish-green fluorescence. As dead cells lack cytoplasmic esterases, fluorescence is demonstrated exclusively in live cells, making the probe useful for determining cell viability. Calcein-AM exhibits low cytotoxicity and does not significantly affect cellular functions such as proliferation or chemotaxis. Viability assays using calcein correlate well with other assays, such as 51Cr-release. Wavelength Maxima: Excitation 495nm, Emission 515nm

Formula: C₄₆H₄₆N₂O₂₃
MW: 994.86 g/mol
Storage Temperature: Freezer
MDL Number: MFCD05861516
CAS Number: 148504-34-1
https://ca.vwr.com/stibo/web/std.lang.all/15/85/45401585.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR