PROTEOSTAT® Thermal Shift Stability Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51027-K400 ENZ-51027-K100
89165-918EA 1000 CAD
89165-918 76002-532
PROTEOSTAT® Thermal Shift Stability Assay Kit, Enzo Life Sciences
Assays Protein Assays
This kit has been designed for monitoring protein stability under systematic thermal stress conditions.
  • Accelerated screening for protein stability as a function of pH, ionic strength, and concentration
  • Can detect ligand binding, even without a prior knowledge of the protein’s function or ligand binding site
  • Provides a convenient, complementary orthogonal method for cross-validation of instrumentation-intensive techniques
The aggregation temperature is an indicator of protein stability and can be used to optimize conditions that minimize protein aggregation as well as to identify ligands that bind and confer structural stability to a protein of interest. Conditions that increase the aggregation temperature, increase the stability of the protein.
Delivery information: Kit includes PROTEOSTAT® TS Detection Reagent, β-Lactoglobulin control, 10X Assay Buffer.