GFP-CERTIFIED® FLUOFORTE® Calcium Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
The GFP-Certified™ FluoForte™ Calcium Assay kit provides a homogeneous fluorescence-based assay for detecting intracellular calcium mobilization across a broad spectrum of biological targets.
Relative to other commercially available dyes, the GFP-Certified™ FluoForte™ dye is the brightest and most sensitive fluorescent calcium indicator. The kit provides a homogeneous mix-and-read, no-wash calcium mobilization assay. The homogenous cell-based assay for calcium offers fewer steps, lower variability and an easier protocol for adherent and non-adherent cell lines. In addition, it requires neither a washing step, nor exogenous addition of a quencher dye, which could adversely effect receptor-ligand interaction kinetics.
Delivery information: Starter kit includes Reagent A (lyophilized GFP-CERTIFIED® FLUOFORTE® dye), Reagent B (dye efflux inhibitor), Reagent C (Hanks’ buffer with 20mM HEPES).
High-throughput kit includes Reagent A (lyophilized GFP-CERTIFIED® FLUOFORTE® dye), Reagent B (dye efflux inhibitor).