Bio-16-ddUTP . tetralithium salt, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42813
89165-830EA 546 CAD
89165-830
Bio-16-ddUTP . tetralithium salt, Enzo Life Sciences
Nucleic Acid Reagents Nucleotides
Bio-16-ddUTP (Biotin-16-2’,3’-dideoxyuridine-5’-triphosphate) can be used for 3’-end labeling of single-stranded or double-stranded DNA. It is especially useful as a substrate for terminal transferase for the 3’-terminal addition of a single Bio-16-ddUMP. Biotin-labeled oligonucleotides produced by this reaction are suitable as hybridization probes.
Biotin-labeled cDNA produced by this reaction can be used in nucleic acid array assays. Biotin-labeled DNA in hybridization assays can be detected by a reporter molecule linked to streptavidin, avidin, or and anti-biotin antibody. Such a complex can be detected directly, e.g. by the excitation of a fluorophore conjugated to streptavidin, or indirectly, e.g. using an enzyme conjugate that can produce an insoluble colored precipitate. Bio-16-ddUTP is also a substrate for Taq DNA Polymerase and other DNA polymerases.