Red 650 dUTP, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42522
89165-760EA 651.01 CAD
89165-760
Red 650 dUTP, Enzo Life Sciences
Nucleic Acid Reagents Nucleotides
Red 650 [Cyanine-5E] dUTP is a cyanine-5 derivative with enhanced spectral properties. This nucleotide analog can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase.
Fluorescently labelled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, cDNA labelling and 3'-end labelling.Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Red 650 dUTP can also be used for multicolor fluorescence labelling.