Cyanine 5-UTP (enhanced), Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42506
89165-756EA 651.01 CAD
89165-756
Cyanine 5-UTP (enhanced), Enzo Life Sciences
Nucleic Acid Reagents Nucleotides
Cyanine 5-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP.