BIO-PROBE® Blur 8 Probe, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-40849
89165-736EA 585 CAD
89165-736
BIO-PROBE® Blur 8 Probe, Enzo Life Sciences
Nucleic Acid Reagents Primers and Probes
The Blur 8 BioProbe® labeled probe is prepared by nick translation of a clone of the human alu repeat sequence.
Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).