N-Arachidonoyl-serotonin ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-VR106-0010 BML-VR106-0050
89165-508EA 176.96 CAD
89165-508 89165-510
N-Arachidonoyl-serotonin ≥98% (by TLC)
N-Arachidonoyl-serotonin

TRPV1 antagonist


Unnatural fatty acid-neurotransmitter conjugate. Potent TRPV1 antagonist (IC50=40nM). Inhibits fatty acid amide hydrolase (FAAH) (IC50=5.6mM). Enhances levels of anandamide and 2-AG in vivo. Displays analgesic activity in both acute and chronic peripheral pain models but shows little efficacy in an inflammatory bowel disease model.

Formula: C₃₀H₄₂N₂O₂
MDL Number: MFCD02179189
CAS Number: 187947-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e31dac2801e7ac0bafc0e36765ae218eb38a6bd8&vid=1a900e10b696c5cd9bf7eb00ec52bd2561ca1ae9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR