SB-366791 ≥98% (by TLC)
Supplier: Enzo Life Sciences
|
Warning
|
Synonyms:
4-Chloro-N-(3-methoxyphenyl)cinnamide, 3-(4-Chlorophenyl)-N-(3-methoxyphenyl)acrylamide, N-(3-Methoxyphenyl)-4-chlorocinnamide
BML-VR105-0005
BML-VR105-0025
89165-504EA
140.62
CAD
89165-504
89165-506
SB-366791 ≥98% (by TLC)
SB-366791
A potent and selective vanilloid receptor (TRPV1) antagonist. An effective antagonist of hTRPV1 when stimulated by capsaicin, acid, or noxious heat.
- Formulation: Beige solid.
Soluble in DMSO (25 mg/ml) or 100% ethanol (10 mg/ml with warming).
Formula:
C₁₆H₁₄ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD01033818 CAS Number: 472981-92-3 |
|
Specification Test Results
Purity | ≥98% (TLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...