Docetaxel ≥98% (by HPLC)
Supplier: Enzo Life Sciences
|
Danger
|
BML-T129-0005
BML-T129-0025
BML-T129-0250
89165-014EA
165.9
CAD
89165-014
89165-016
89165-018
Docetaxel ≥98% (by HPLC)
Docetaxel
Docetaxel is taxane-type antineoplastic agent structurally similar to taxol.
- Formulation: White solid.
It reversibly binds to polymerised tubulin inhibiting depolymerisation. It induces apoptosis in a large number of tumor cells and is a clinically important anticancer agent. Stabilises microtubules.
Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml).
Synonym: Taxotere
Formula:
C₄₃H₅₃NO₁₄ Storage Temperature: Freezer |
MDL Number:
MFCD00800737 CAS Number: 114977-28-5 |
|
Specification Test Results
Purity | ≥98% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...