Wiskostatin ≥98%
Supplier: Enzo Life Sciences
BML-T126-0001
BML-T126-0005
89165-008EA
137.46
CAD
89165-008
89165-010
Wiskostatin ≥98%
Wiskostatin
Blocks actin filament assembly
A selective, reversible inhibitor of N-WASP (neural Wiskott-Aldrich syndrome protein) that selectively blocks actin filament assembly. Appears to bind to N-WASP, stabilize the autoinhibited conformation and prevent the activation of Arp2/3 (actin-related protein 2/3) complex.
Formula:
C₁₇H₁₈Br₂N₂O |
MDL Number:
MFCD00218393 CAS Number: 253449-04-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...