Vincristine sulfate ≥98%

Supplier: Enzo Life Sciences

BML-T117-0005 BML-T117-0025
89164-990EA 246.48 CAD
89164-990 89164-992
Vincristine sulfate ≥98%
Vincristine sulphate

Microtubule inhibitor


Plant alkaloid that arrests the cell cycle in G2/M phase by blocking mitotic spindle formation. Depolymerizes microtubules and blocks binding of tubulin to microtubule proteins. Causes aberrant tubulin polymerization. Induces apoptosis. Triggers Raf-1 activation, phosphorylation of Bcl-2 family proteins and induction of p53 expression. Cancer chemotherapeutic agent. Posttranscriptionally downregulates HIF-1α expression.

Formula: C₄₆H₅₈N₄O₁₄S
MDL Number: MFCD00084729
CAS Number: 2068-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3751449e4a5845753af6a8c3fd27b1a720b75321&vid=dc1d6c45f09c07ca2ffeb35558a455552f36b543&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR