Parthenolide ≥97% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 4,5alpha-Epoxy-6beta-hydroxygermacra-1(10),11(13)-dien-12-oic Acid gamma-Lactone, (3aS,9aR,10aR,10bS,E)-6,9a-Dimethyl-3-methylene-3a,4,5,8,9,9a,10a,10b-octahydrooxireno[2',3':9,10]cyclodeca[1,2-b]furan-2(3H)-one

BML-T113-0050 BML-T113-0250
89164-978EA 162.74 CAD
89164-978 89164-980
Parthenolide ≥97% (by TLC)
Parthenolide

HDAC inhibitor


Sesquiterpene lactone. Widely used as an herbal remedy for arthritis and migraine. Has anti-inflammatory, antisecretory and spasmolytic activity. Specifically inhibits activation of NF-kappaB by preventing the degradation of IkappaBalpha and IkappaBbeta. Inhibits activation of MAP kinase (MAPK/ERK) and generation of leukotriene B4 and thromboxane B2. Potent anticancer agent. Induces apoptosis in various cancer cell lines. Specifically inhibits histone deacetylase 1 (HDAC1) without affecting other class I/II HDACs. Inhibits tubulin carboxypeptidase (TCP) activity.


Isolated from feverfew (Chrysanthemum parthenium) leaves.

Formula: C₁₅H₂₀O₃
MW: 248.32 g/mol
MDL Number: MFCD00134592
CAS Number: 20554-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR