Paclitaxel ≥98%

Supplier: Enzo Life Sciences

BML-T104-0005 BML-T104-0025 BML-T104-0250
89164-960EA 140.62 CAD
89164-960 89164-962 89164-964
Paclitaxel ≥98%
Paclitaxel

Ultra-pure; stabilizes microtubules


Antitumor agent. Stabilizes microtubules and lowers the critical concentration for tubulin polymerization. Reversibly binds to polymerized tubulin. Inhibits protein isoprenylation and induces apoptosis in PC-3 human prostate cancer cells. The checkpoint of mitotic spindle assembly, aberrant activation of cyclin-dependent kinases (CKDs) and the c-Jun N-terminal kinase/stress activated protein kinase (JNK/SAPK) are involved in Paclitaxel-induced apoptosis.


Isolated from the bark of the Pacific Yew tree (Taxus brevifolia)

Formula: C₄₇H₅₁NO₁₄
MDL Number: MFCD00869953
CAS Number: 33069-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26dcc87c6ca0f0093d7466f748f1a54718456cfd&vid=6802cd4f4bd14df43040bc1abc8bfc76b2dc2ec2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR