Arachidonoyl-AMC ≥99%

Supplier: Enzo Life Sciences

BML-ST504-0005 BML-ST504-0025
89164-888EA 188.02 CAD
89164-888 89164-890
Arachidonoyl-AMC ≥99%
Arachidonoyl-AMC

FAAH substrate.


  • Formulation: Off-white solid.


Fluorogenic substrate for fatty acid amide hydrolase (FAAH). This substrate is the basis for a simple, selective and sensitive assay for FAAH which is amenable to high-throughput screening.


Soluble in DMSO (>25 mg/ml).

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea8145f99e0dcf1eeb6f64901a4fe55210a72c0b&vid=1eecb728e3f8b07aa6fda7543ad36db9d80970e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR