Phosphatidylbutanol ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-ST401-0010
BML-ST401-0050
89164-854EA
608.3
CAD
89164-854
89164-856
Phosphatidylbutanol ≥98% (by TLC)
Phosphatidylbutanol
Phosphatidylbutanol is a useful chromatographic standard for PLD assay.
- Formulation: Viscous oil.
Soluble in chloroform.
|
Specification Test Results
Purity | ≥98% (TLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...