Phosphatidylethanol ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol, PEth-16:0/18:1

BML-ST400-0010 BML-ST400-0050
89164-850EA 237 CAD
89164-850 89164-852
Phosphatidylethanol ≥98% (by TLC)
Phosphatidylethanol

Ultra-pure


  • Formulation: Viscous oil or waxy solid.


In the presence of ethanol, phospholipase D converts phosphatidylcholine to phosphatidylethanol. This conversion is the basis of a sensitive and selective assay for PLD activity in intact cells. Phosphatidylethanol is a useful chromatographic standard for this assay.


Soluble in chloroform.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3cce0484ac9e439a7567882c668939a10aaac2c&vid=da73a9d470718089ce8b577cd7e3b9945528dd91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR