U-73343 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 1-[6-[[(17b)-3-methoxyestra-1,3,5(10)-trien-17-yl]amino]hexyl]-2,5-pyrrolidinedione

BML-ST392-0005 BML-ST392-0025
89164-846EA 237 CAD
89164-846 89164-848
U-73343 ≥98% (by HPLC)
U-73343

Inactive analog of U-73122 which only slightly inhibits phospholipase C. May be used as a negative control.


  • Phospholipase inhibitor


White to off-white solid.


Soluble in methylene chloride or chloroform; slightly soluble in DMSO (2mg/ml) or 100% ethanol (1mg/ml); sparingly soluble in aqueous solutions.

Formula: C₂₉H₄₂N₂O₃
MW: 466.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00211221
CAS Number: 142878-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db4bb882c407085c163ec076b8ce67d7615ce0bd&vid=6ca940de98bcd96e0907ff1440c443a5438303eb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR