U-73122 ≥99% (by TLC)
Supplier: Enzo Life Sciences
Inhibitor of PLC activation
Inhibits agonist induced phospholipase C (PLC) activation (IC50=1-2.1 µM) in human platelets. It also inhibits agonist induced down-regulation of muscarinic receptors in SK-N-SH neuroblastoma cells. It is a useful tool to investigate receptor-mediated PI turnover in signal transduction. It is a potent inhibitor of human neutrophil adhesion to biological surfaces as well as adhesion-dependent granule exocytosis and oxidative burst. Inhibits 5-lipoxygenase in vivo and in vitro (IC50 = 30nM for recombinant enzyme and 2.4µM for PMNL homogenates).
Formula:
C₂₉H₄₀N₂O₃ |
MDL Number:
MFCD00893825 CAS Number: 112648-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...