Fingolimod hydrochloride ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2-Amino-2-(4-octylphenethyl)propane-1,3-diol hydrochloride, FTY-720 hydrochloride

BML-SL233-0005 BML-SL233-0025
89164-676EA 151.68 CAD
89164-676 89164-678
Fingolimod hydrochloride ≥98% (by HPLC)
Fingolimod hydrochloride

Novel immune modulator that alters migration and homing of lymphocytes. Acts as a potent agonist of sphingosine 1-phosphate receptors (S1P) after phosphorylation by sphingosine kinase. Also inhibits cytosolic phospholipase A2 independently of sphingosine 1-phosphate receptors.


  • S1P receptor ligand


White solid.


Soluble in DMSO (10mg/ml), 100% ethanol or dimethyl formamide.

Formula: C₁₉H₃₄ClNO₂
MW: 343.936 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00939512
CAS Number: 162359-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR