α-Galactosylceramide (synthetic) ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: alpha-GalCer, alpha-Galactosylceramide, N-((2S,3S,4R)-3,4-Dihydroxy-1-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)octadecan-2-yl)hexacosanamide, KRN7000

BML-SL232-0100 BML-SL232-1000
89164-672EA 627.26 CAD
89164-672 89164-674
α-Galactosylceramide (synthetic) ≥99% (by HPLC)
α-Galactosylceramide

MHC ligand.


  • Formulation: White solid.


KRN7000 is a synthetic analog of α-galactosylceramide and the marine natural product agelasphin. It is a specific ligand for human and mouse NKT cells and remains the best studied ligand of the lipid-binding MHC class I-like protein CD1d. It protects against LPS-induced shock and displays potent antitumor activity in various in vivo models.


KRN7000 is inherently an extremely hydrophobic molecule, therefore, almost all the methods for solubilizing this material in primarily aqueous media will contain at least some detergent. For cell culture or other in vivo models, it is recommended that the KRN7000 be initially dissolved in a 2:1 mixture of chloroform and methanol.

Formula: C₅₀H₉₉NO₉
MW: 858.34 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00939559
CAS Number: 158021-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7334f05253383bf78b11488c03d8c2b4ce01235a&vid=d7505aa4e2fb9b4fc828461371cce30ab4b9f384&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR