N-Butyldeoxynojirimycin hydrochloride ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-SL230-0005 BML-SL230-0025
89164-664EA 259.12 CAD
89164-664 89164-666
N-Butyldeoxynojirimycin hydrochloride ≥98% (by TLC)
N-Butyldeoxynojirimycin hydrochloride

A novel inhibitor of glucosylceramide synthase as well as glucosidase I and II


  • White to off-white solid


Inhibits glucosylceramide synthase without ceramide accumulation and cell death. It is a potent inhibitor of human immunodeficiency virus (HIV) replication and syncytium formation in vitro. Restores chloride current in epithelial CF cells.


Soluble in water

Formula: C₁₀H₂₁NO₄·HCl
Storage Temperature: Freezer
MDL Number: MFCD00269940
CAS Number: 210110-90-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99f53856b1f856dd4f261ce99cc402bb08af220e&vid=ccad14a0bf2bf2a334dbb9af4e0359fbc2a9ed1b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR