D-erythro MAPP ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-SL221-0005 BML-SL221-0025
89164-646EA 137.46 CAD
89164-646 89164-648
D-erythro MAPP ≥98% (by TLC)
D-erythro MAPP

Alkaline ceramidase inhibitor


A cell-permeable and specific inhibitor of alkaline ceramidase, which can raise intracellular ceramide levels by blocking the hydrolysis of ceramide to sphingosine and free fatty acid. Enhances the apoptotic response to ionizing radiation by enhancing the ceramide signal.

Formula: C₂₃H₃₉NO₂
MDL Number: MFCD01076527
CAS Number: 143492-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03bee86f1e512f352ea87307db78149cb516447d&vid=a6552b01de010423cfa57fd681244ee8f7033ac8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR