Fumonisin B1 ≥98%

Supplier: Enzo Life Sciences

BML-SL220-0001 BML-SL220-0005
89164-642EA 184.86 CAD
89164-642 89164-644
Fumonisin B1 ≥98%
Fumonisin B1

Ceramide synthase inhibitor


A fungal metabolite produced by Fusarium moniliforme. Mycotoxin. Inhibits sphinganine N-acyl-transferase(ceramide synthase)(IC50=0.1µM). Inhibits de novo sphingolipid biosynthesis (IC50=0.7µM for sphingomyelin labeling by [3H]sphinganine, cultured cerebellar neurons). Inhibits growth of axons in cultured hippocampal neurons by blocking glycosphingolipid synthesis. Induces DNA damage in the liver and cancer. Induces apoptosis. Activates Akt (protein kinase B; PKB), which leads to increased survival, inhibition of GSK-3β activity and post-translational stabilization of cyclin D1.

Formula: C₃₄H₅₉NO₁₅
MW: 721.838 g/mol
MDL Number: MFCD00133349
CAS Number: 116355-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38ebdb7a105882b0e23e5656b07dbc44dc112319&vid=935600c5fa936d8dd0fa2d0787ac2f2a72dbc64d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR