Fumonisin B2 ≥80%

Supplier: Enzo Life Sciences

BML-SL219-0001 BML-SL219-0005
89164-638EA 483.48 CAD
89164-638 89164-640
Fumonisin B2 ≥80%
Fumonisin B2

Ceramide synthase inhibitor


Mycotoxin, structurally similar to fumonisin B1 (Prod. No. BML-SL220). Found as contaminant mostly in corn silage; was detected recently for the first time in the industrially important Aspergillus niger. Induces cancer. Inducer of apoptosis. Inhibits sphinganine N-acyltransferase (ceramide synthase) at IC50 = 0.1µM. Inhibits de novo sphingolipid biosynthesis.

Formula: C₃₄H₅₉NO₁₄
MDL Number: MFCD00133350
CAS Number: 116355-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR