DL-threo-1-Phenyl-2-palmitoylamino-3-morpholino-1-propanol ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: DL-PPMP

BML-SL215-0005 BML-SL215-0025
89164-634EA 115.34 CAD
89164-634 89164-636
DL-threo-1-Phenyl-2-palmitoylamino-3-morpholino-1-propanol ≥98% (by TLC)
DL-threo-1-Phenyl-2-palmitoylamino-3-morpholino-1-propanol

Inhibits glucosylceramide synthetase. More effective than PDMP in intact cells.


  • Glucosylceramide synthetase inhibitor


White-pink solid.


Soluble in DMSO (10mg/ml).

Formula: C₂₉H₅₀N₂O₃·HCl
Storage Temperature: Freezer
CAS Number: 149022-18-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43481e91a70316b05ef52a0bbd8f0e9f90a675cb&vid=b10255ec179227f27bfc42213b326e6122fd80e7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR