D,L-threo-PDMP hydrochloride ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: DL-threo-1-Phenyl-2-decanoylamino-3-morpholino-1-propanol hydrochloride, D,L-threo-PDMP hydrochloride, 1-Phenyl-2-decanoylamino-3-morpholino-1-propanol HCl

BML-SL210-0010 BML-SL210-0050
89164-630EA 116.92 CAD
89164-630 89164-632
D,L-threo-PDMP hydrochloride ≥98% (by TLC)
D,L-threo-PDMP hydrochloride

Blocks glucosylation of ceramide by inhibiting UDP-glucose:ceramide glucosyltransferase (glucosylceramide synthetase). Inhibits the expression of cell surface glycolipid antigens and inhibits the growth of cultured rabbit skin fibroblasts. Possesses antitumor activity and inhibits Lewis lung carcinoma cell metastasis in vitro. Useful tool for studying the effects of cellular glycosphingolipid depletion. Enhances the apoptotic response to ionizing radiation by enhancing the ceramide signal.


  • Glucosylceramide synthetase inhibitor


White to off-white solid.


25mg/ml soluble in 100% ethanol, DMSO or water (50°C, vortex or sonicate).

Formula: C₂₃H₃₈N₂O₃·HCl
Storage Temperature: Freezer
MDL Number: MFCD00236490
CAS Number: 80938-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feeb4ff60f78e4225484e7229b76e9b32250a07d&vid=938ee1e14b4cff6c06da680ddd364e4ec244d413&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR