Sphingomyelin ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: N-Acyl-4-sphingenyl-1-O-phosphorylcholine, N-Acyl-D-sphingosine-1-phosphocholine

BML-SL135-0100 BML-SL135-0500
89164-602EA 237 CAD
89164-602 89164-604
Sphingomyelin ≥98% (by TLC)
Sphingomyelin

Purified from bovine brain. Contains primarily stearic and nervoic acids. Precursor to ceramide second messengers via the action of sphingomyelinase.


Soluble in chloroform, ethanol or methanol.

Formula: C₄₁H₈₃N₂O₆P
Storage Temperature: Freezer
MDL Number: MFCD00132361
CAS Number: 85187-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a40d0aa5e8bdaec74c26f8a34c53d4201f9bc93&vid=a1647083bb77ddf3b03dc7b78faa8dfcd493f66f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR