N-Palmitoyl ceramide ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: D-erythro-Sphingosine, N-Palmitoyl-, N-Palmitoyl-D-erythro-sphingosine, C16 Ceramide, N-Hexadecanoyl-D-erythro-sphingosine

BML-SL115-0005 BML-SL115-0025
89164-592EA 137.46 CAD
89164-592 89164-594
N-Palmitoyl ceramide ≥98% (by TLC)
N-Palmitoyl ceramide

Endogenous ceramide.


An abundant molecular species of endogenous ceramide. It induces phosphorylation of epidermal growth factor receptor on Thr⁶⁶⁹ in A-431 cells by stimulation of ceramide-activated protein kinase and activates NF-κB at concentrations as low as 50 nM. Activates PKCζ.


Soluble in 100% ethanol (5 mg/ml warm).


Synonym: N-Palmitoyl-D-erythro-sphingosine, N-Hexadecanoyl-D-erythro-sphingosine, N-Palmitoyl ceramide

Formula: C₃₄H₆₇NO₃
Storage Temperature: Freezer
MDL Number: MFCD00871748
CAS Number: 24696-26-2

Order Now

Specification Test Results

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR