N,N-Dimethylsphingosine, D-erythro ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-SL105-0005 BML-SL105-0025
89164-572EA 148.52 CAD
89164-572 89164-574
N,N-Dimethylsphingosine, D-erythro ≥98% (by TLC)
N,N-Dimethylsphingosine, D-erythro
Sphingosine kinase inhibitor
Potent and specific inhibitor of sphingosine kinase (IC50=5µM) which blocks conversion of sphingosine to sphingosine-1-phosphate (Prod. No. BML-SL140). Inhibitor of protein kinase C (PKC) which also stimulates Src-kinase. May affect expression of cell surface selectins, which in turn mediate leukocyte or tumor cell adhesion to endothelial cells and platelets. Induces apoptosis. Stereospecifically induces EGF receptor autophosphorylation producing EGF-like activity in vitro in the absence of EGF. Produced endogenously via ceramide catabolism. May be a target for neuropathic pain therapy.
Synthetic.
CAS Number: 119567-63-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21d0b9fc6a7cf9abb2042276457cca892de2dfe1&vid=cccc773ff67e807fa679cf991422c0248a1ba1c9&prid=web&clid=SAPDAM