C2 Ceramide ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-SL100-0005
BML-SL100-0025
89164-564EA
132.72
CAD
89164-564
89164-566
C2 Ceramide ≥98% (by TLC)
C2 Ceramide
Cell-permeable ceramide analog
N-Acetylsphingosine is a biologically active, cell-permeable ceramide analog. It inhibits cell proliferation and induces monocytic differentiation of HL-60 cells and induces apoptosis. It stimulates protein phosphatase 2A4, activates MAP kinase and SAP kinase and induces PKCδ and ε translocation. Physiological levels of C2 ceramide, produced by the action of a PAF:sphingosine CoA-independent transacetylase, have been detected in HL-60 cells.
Formula:
C₂₀H₃₉NO₃ |
MDL Number:
MFCD00153903 CAS Number: 3102-57-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...