Anti-Insulin Receptor Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA392-0020 BML-SA392-0100
89162-990EA 290.72 CAD
89162-990 89162-992
Anti-Insulin Receptor Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes human and mouse insulin receptor/IGF-1R phosphorylated at Tyr1158/1162/1163.

Type: Primary
Antigen: INSULIN RECEPTOR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR