Anti-COX-2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA237-0025 BML-SA237-0100
89162-760EA 268.6 CAD
89162-760 89162-762
Anti-COX-2 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and sheep COX-2. Does not cross-react with COX-1.

Type: Primary
Antigen: COX-2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR