Pregnenolone-16-α-carbonitrile ≥99% (by HPLC)

Supplier: Enzo Life Sciences

BML-S620-0020 BML-S620-0100
89162-606EA 88.48 CAD
89162-606 89162-608
Pregnenolone-16-α-carbonitrile ≥99% (by HPLC)
Pregnenolone-16-alpha-carbonitrile

Cytochrome P450 inducer.


  • Formulation: White to off-white solid.


Activates transcription of the PCN1 gene. SXR/PXR Ligand.


Soluble in DMF (5 mg/ml) or acetone (1 mg/ml).


Synonym: U-14975

Formula: C₂₂H₃₁NO₂
Storage Temperature: Freezer
CAS Number: 1434-54-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8e434ba0e21bba7ca79ec0fddc242bf6ecb236e&vid=ecef65d17d000e8e00064b0c2a3c9b617102e11f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR