Mifepristone ≥99%

Supplier: Enzo Life Sciences
Danger

BML-S510-0025 BML-S510-0100
89162-590EA 172.22 CAD
89162-590 89162-592
Mifepristone ≥99%
Mifepristone

Progesterone receptor ligand.


  • Formulation: Yellow solid.


Progesterone receptor antagonist.


Soluble in methanol or DMSO.


Synonym: RU 486

Formula: C₂₉H₃₅NO₂
Storage Temperature: Freezer
CAS Number: 84371-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=716871e6caa42d45499b47346f5d2417dc14885a&vid=b121b4e51ffa2524223fc91238aa9b18cfcc68cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99% (Assay)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR