Kifunensine 98 TLC_ASSAY_METHOD

Supplier: Enzo Life Sciences

BML-S114-0001 BML-S114-0005
89162-572EA 224.36 CAD
89162-572 89162-574
Kifunensine 98 TLC_ASSAY_METHOD
Kifunensine

Inhibits α-mannosidase. Inhibits asparagine-linked oligosaccharide processing. Immunomodulator.


  • Mannosidase inhibitor


White to off-white solid.


Soluble in water (25mg/ml).

Formula: C₈H₁₂N₂O₆
MW: 232.192 g/mol
Storage Temperature: Freezer
CAS Number: 109944-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91d671159b983a2b9b447ff141e04a99a4a9566d&vid=7f485c19a1e14251b738b9f4a42d53cc3a28b823&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR