Swainsonine ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-S112-0001 BML-S112-0005
89162-564EA 338.12 CAD
89162-564 89162-566
Swainsonine ≥98% (by HPLC)
Swainsonine

Reversibly inhibits the active site of lysosomal α-mannosidase. Also inhibits the growth of tumors and prevents metastasis in murine models. Could inhibit cell proliferation in vitro and the growth of human gastric carcinoma in vivo. Blocks the processing of high mannose oligosaccharides to form complex type oligosaccharides.^Immunomodulator.


  • Lysosomal alpha-mannosidase inhibitor


White to off-white solid.


Soluble in 100% ethanol, DMSO, methanol or water (10mg/ml).

Formula: C₈H₁₅NO₃
Storage Temperature: Refrigerator
CAS Number: 72741-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26dc088d6a42437db6a55f8ee2860b37acc9548e&vid=294ebaf2b8358616561fd5d2c8343a6ab413dedd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR