Seratrodast ≥98%

Supplier: Enzo Life Sciences
Warning

Synonyms: 7-Phenyl-7-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)heptanoic acid

BML-RA119-0010 BML-RA119-0050
89162-528EA 194.34 CAD
89162-528 89162-530
Seratrodast ≥98%
Seratrodast

Thromboxane receptor ligand.


  • Formulation: Yellow-orange solid.


Seratrodast is a potent and selective thromboxane A₂ receptor (TP) antagonist with antioxidant properties.


Soluble in DMSO (12 mg/ml); Ethanol (10 mg/ml)

Formula: C₂₂H₂₆O₄
Storage Temperature: Ambient
MDL Number: MFCD00875701
CAS Number: 112665-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=facd2bb44ca138f65fcd31ac0386646c8958ff67&vid=9a23b76c3fd6bdf368d2a5fc9d5dee6bd396decf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR