SC-51322 ≥98%
Supplier: Enzo Life Sciences
Synonyms:
3-chloro-N'-[3-(furan-2-ylmethylsulfanyl)propanoyl]-6H-benzo[b][1,4]benzoxazepine-5-carbohydrazide, 3-chloro-N'-[3-(furan-2-ylmethylsulphanyl)propanoyl]-6H-benzo[b][1,4]benzoxazepine-5-carbohydrazide
BML-RA112-0005
BML-RA112-0025
89162-500EA
172.22
CAD
89162-500
89162-502
SC-51322 ≥98%
SC-51322
Prostanoid receptor inhibitor
A potent and selective prostaglandin E2 antagonist (pA2=8.1). Displays in vivo analgesic activity in the mouse (ED50=0.9 mg/kg).
Formula:
C₂₂H₂₀ClN₃O₄S |
MDL Number:
MFCD00916246 CAS Number: 146032-79-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...