MK-571 sodium salt ≥90% (by HPLC)

Supplier: Enzo Life Sciences
Warning

BML-RA109-0005 BML-RA109-0025
89162-492EA 140.62 CAD
89162-492 89162-494
MK-571 sodium salt ≥90% (by HPLC)
MK-571 sodium salt

A potent, selective, orally active leukotriene D₄ antagonist. Ki=2.1 nM for inhibition of [³H]-LTD₄ binding to human lung membranes. Competitively antagonizes contractions of guinea pig trachea and ileum and human trachea induced by LTD₄. Blocks LTD₄ and ascaris-induced bronchoconstriction in conscious squirrel monkeys.


  • Formulation: Off-white solid.


Soluble in water (20 mg/ml), aqueous buffers, 100% ethanol (10 mg/ml) or DMSO.


Synonym: L660,711

Formula: C₂₆H₂₆ClN₂O₃S₂·Na
Storage Temperature: Freezer
MDL Number: MFCD11114392
CAS Number: 115103-85-0

Order Now

Specification Test Results

Purity ≥90% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR