BML-111 ≥99%
Supplier: Enzo Life Sciences
Synonyms:
5(S),6(R),7-Trihydroxyheptanoic acid methyl ester
BML-RA108-0001
BML-RA108-0005
89162-488EA
140.62
CAD
89162-488
89162-490
BML-111 ≥99%
BML-111
Potent lipoxin A₄ agonist, equiactive as LXA₄ in inhibition of LTB₄ induced PMNL chemotaxis (IC₅₀ = 5 nM).
- Formulation: White solid.
Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml).
Synonym: 5(S),6(R),7-Trihydroxyheptanoic acid methyl ester
Formula:
C₈H₁₆O₅ MW: 192.2 g/mol Storage Temperature: Ambient |
CAS Number:
78606-80-1 |
Specification Test Results
Purity | ≥99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...