Anti-HERP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW9705-0025 PW9705-0100
89162-412EA 325.48 CAD
89162-412 89162-414
Anti-HERP Rabbit Polyclonal Antibody
Antibodies
Homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domain member 1 protein (HERP) is a membrane-associated ER protein strongly up-regulated by inducers of ER stress.

  • For Immunoprecipitation, Western Blot

Recognizes human HERP.

Type: Primary
Antigen: HERP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR