Anti-Proteasome 20S Beta7 Subunit Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW9150-0025 PW9150-0100
89162-304EA 254.38 CAD
89162-304 89162-306
Anti-Proteasome 20S Beta7 Subunit Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunoprecipitation, Western Blot

Recognizes human proteasome 20S (subunit beta7).

Type: Primary
Antigen: PROTEASOME 20S BETA7 SUBUNIT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR